Learn More
(+)-Quisqualic acid, 99+%, Thermo Scientific Chemicals
Description
Quisqualic acid is used to differentiate between mGluR-1 and mGluR-4. L-Quisqualic acid is an activator of GluR.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Quisqualic acid is used to differentiate between mGluR-1 and mGluR-4. L-Quisqualic acid is an activator of GluR.
Solubility
Soluble in ethanol, water. Insoluble in organic solvents.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 185°C to 187°C |
| Color | White |
| Quantity | 10 mg |
| Beilstein | 1078734 |
| Merck Index | 14,8085 |
| Solubility Information | Soluble in ethanol,water. Insoluble in organic solvents. |
| Formula Weight | 189.13 |
| Percent Purity | ≥99% |
| Physical Form | Powder |
| Chemical Name or Material | (+)-Quisqualic acid |
Safety and Handling
| Hazard Category | H302+H312+H332 |
| Hazard Statement | GHS H Statement H302-H312-H332 Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.