Learn More
AdipoGen Quizalofop-P
SDP
Supplier:  AdipoGen CDXQ0009M100
Encompass_Preferred
Chemical. CAS 94051-08-8. Formula C17H13CIN2O4. MW 344.75. Synthetic. Aryloxyphenoxypropionic herbicide. Used for the post-emergence control of annual and perennial grass weeds in various field crops. Mode of action is through inhibition of acetyl CoA carboxylase ACCase. Compound can be used as analytical reference material.
Catalog No. 50-256-0165