Learn More
Medchemexpress LLC Quizalofop-p-ethyl ((R)-Quizalofop ethyl) | 100646-51-3 | 99.1% | 200 MG
SDP
Catalog No. 501970174
Encompass_Preferred
Quizalofop-P-ethyl is a low-toxicity herbicide, characterized as a highly selective, new type of post-emergence herbicide for dryland crops. It demonstrates high selectivity between gramineous weeds and dicotyledonous crops, offering effective control against gramineous weeds in broadleaf crop fields.
  • This compound inhibits the growth of a specific aquatic microorganism and reduces its chlorophyll and protein content.
  • It impacts photosynthesis by suppressing the expression of photosynthesis-related genes.
  • The compound encourages the expression of a gene related to microcystin, stimulating the synthesis and release of microcystin, which can lead to water environment pollution.
  • It also increases MDA content in the aquatic microorganism and enhances the activities of SOD and CAT, inducing oxidative stress.
Catalog No. 50-197-0174 Supplier Medchemexpress LLC Supplier No. HYB1950200MG
Quizalofop-P-ethyl is a low-toxicity herbicide, characterized as a highly selective, new type of post-emergence herbicide for dryland crops. It demonstrates high selectivity between gramineous weeds and dicotyledonous crops, offering effective control against gramineous weeds in broadleaf crop fields.
  • This compound inhibits the growth of a specific aquatic microorganism and reduces its chlorophyll and protein content.
  • It impacts photosynthesis by suppressing the expression of photosynthesis-related genes.
  • The compound encourages the expression of a gene related to microcystin, stimulating the synthesis and release of microcystin, which can lead to water environment pollution.
  • It also increases MDA content in the aquatic microorganism and enhances the activities of SOD and CAT, inducing oxidative stress.