Learn More
Medchemexpress LLC HY-13001 100mg , Quizartinib CAS:950769-58-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13001100MG
Encompass_Preferred
HY-13001 100mg Quizartinib CAS: 950769-58-1 Quizartinib (AC220) is an orally active, highly selective and potent second-generation type II FLT3 tyrosine kinase inhibitor, with a Kd of 1.6 nM. Quizartinib inhibits wild-type FLT3 and FLT3-ITD autophosphorylation in MV4-11 cells with IC50s of 4.2 and 1.1 nM, respectively. Quizartinib can be linked to the VHL ligand via an optimized linker to form a PROTAC FLT3 degrader. Quizartinib induces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9239