Learn More
Cayman Chemical QuIzartInIb 10mg
SDP
Catalog No. 502979719
Encompass_Preferred
A selective FLT3 inhibitor (Kd = 1.6 nM) that inhibits the proliferation of MV4-11 cells both in vitro (IC50 = 0.56 nM) and in in vivo mouse models of FLT3-ITD acute myeloid leukemia
Catalog No. 50-297-9719 Supplier Cayman Chemical Supplier No. 1798610 MG
A selective FLT3 inhibitor (Kd = 1.6 nM) that inhibits the proliferation of MV4-11 cells both in vitro (IC50 = 0.56 nM) and in in vivo mouse models of FLT3-ITD acute myeloid leukemia