Learn More
Medchemexpress LLC Quizartinib | 950769-58-1 | 99.1% | 200 MG
SDP
Catalog No. 5000339126
Encompass_Preferred
Quizartinib is an orally active, highly selective, and potent second-generation type II FLT3 tyrosine kinase inhibitor. It inhibits wild-type FLT3 and FLT3-ITD autophosphorylation in MV4-11 cells, with IC50s of 4.2 nM and 1.1 nM respectively. This compound can be linked to the VHL ligand via an optimized linker to form a PROTAC FLT3 degrader, and it induces apoptosis. It is intended for research use only.
  • Orally active
  • Highly selective and potent type II FLT3 tyrosine kinase inhibitor
  • Inhibits wild-type FLT3 and FLT3-ITD autophosphorylation
  • Forms PROTAC FLT3 degrader when linked to VHL ligand
  • Induces apoptosis
Catalog No. 50-003-39126 Supplier Medchemexpress LLC Supplier No. HY13001200MG
Quizartinib is an orally active, highly selective, and potent second-generation type II FLT3 tyrosine kinase inhibitor. It inhibits wild-type FLT3 and FLT3-ITD autophosphorylation in MV4-11 cells, with IC50s of 4.2 nM and 1.1 nM respectively. This compound can be linked to the VHL ligand via an optimized linker to form a PROTAC FLT3 degrader, and it induces apoptosis. It is intended for research use only.
  • Orally active
  • Highly selective and potent type II FLT3 tyrosine kinase inhibitor
  • Inhibits wild-type FLT3 and FLT3-ITD autophosphorylation
  • Forms PROTAC FLT3 degrader when linked to VHL ligand
  • Induces apoptosis