Learn More
Medchemexpress LLC Quizartinib dihydrochloride | 1132827-21-4 | 99.8% | 633.59 g·mol⁻1 | C29H34Cl2N6O4S | 5 MG
SDP
Catalog No. 5000221602
Encompass_Preferred
Quizartinib dihydrochloride is the dihydrochloride salt form of quizartinib, a second-generation, highly selective type II FLT3 tyrosine kinase inhibitor used in preclinical oncology research. It demonstrates low-nanomolar potency against FLT3 and FLT3-ITD and is employed in cellular assays and medicinal chemistry applications, including degrader design. The material is supplied at research scale with high reported purity.
  • High potency FLT3 inhibition in the low-nanomolar range.
  • Suitable for in vitro and preclinical AML models.
  • Compatible with PROTAC linker chemistry and conjugation studies.
  • Supplied as the dihydrochloride salt for improved solubility.
  • High purity suitable for biochemical and cellular assays.
  • Available in multiple small research pack sizes.
Catalog No. 50-002-21602 Supplier Medchemexpress LLC Supplier No. HY142175MG
Quizartinib dihydrochloride is the dihydrochloride salt form of quizartinib, a second-generation, highly selective type II FLT3 tyrosine kinase inhibitor used in preclinical oncology research. It demonstrates low-nanomolar potency against FLT3 and FLT3-ITD and is employed in cellular assays and medicinal chemistry applications, including degrader design. The material is supplied at research scale with high reported purity.
  • High potency FLT3 inhibition in the low-nanomolar range.
  • Suitable for in vitro and preclinical AML models.
  • Compatible with PROTAC linker chemistry and conjugation studies.
  • Supplied as the dihydrochloride salt for improved solubility.
  • High purity suitable for biochemical and cellular assays.
  • Available in multiple small research pack sizes.