Learn More
Medchemexpress LLC Quizartinib dihydrochloride | 1132827-21-4 | 99.8% | C29H34Cl2N6O4S | 100 MG
SDP
Catalog No. 5000353307
Encompass_Preferred
Quizartinib dihydrochloride (AC220 dihydrochloride) is the dihydrochloride salt form of Quizartinib. This compound is an orally active, highly selective, and potent second-generation type II FLT3 tyrosine kinase inhibitor, characterized by a Kd of 1.6 nM. It is intended for research use only.
  • Highly selective and potent second-generation type II FLT3 tyrosine kinase inhibitor
  • Effectively inhibits wild-type FLT3 and FLT3-ITD autophosphorylation
  • Can be utilized in PROTAC technology to create FLT3 degraders
  • Capable of inducing apoptosis
  • Provides high purity for reliable research results
  • Presents as a white to off-white solid
Catalog No. 50-003-53307 Supplier Medchemexpress LLC Supplier No. HY14217100MG
Quizartinib dihydrochloride (AC220 dihydrochloride) is the dihydrochloride salt form of Quizartinib. This compound is an orally active, highly selective, and potent second-generation type II FLT3 tyrosine kinase inhibitor, characterized by a Kd of 1.6 nM. It is intended for research use only.
  • Highly selective and potent second-generation type II FLT3 tyrosine kinase inhibitor
  • Effectively inhibits wild-type FLT3 and FLT3-ITD autophosphorylation
  • Can be utilized in PROTAC technology to create FLT3 degraders
  • Capable of inducing apoptosis
  • Provides high purity for reliable research results
  • Presents as a white to off-white solid