Learn More
Medchemexpress LLC Quizartinib dihydrochloride | 1132827-21-4 | 99.8% | C29H34Cl2N6O4S | 50 MG
SDP
Catalog No. 5000353621
Encompass_Preferred
Quizartinib dihydrochloride is an orally active, highly selective, and potent second-generation type II FLT3 tyrosine kinase inhibitor. It inhibits wild-type FLT3 and FLT3-ITD autophosphorylation in MV4-11 cells and can induce apoptosis.
  • Orally active FLT3 tyrosine kinase inhibitor
  • Highly selective and potent
  • Inhibits FLT3 and FLT3-ITD autophosphorylation
  • Can be linked to VHL ligand for PROTAC FLT3 degrader formation
  • Induces apoptosis
Catalog No. 50-003-53621 Supplier Medchemexpress LLC Supplier No. HY1421750MG
Quizartinib dihydrochloride is an orally active, highly selective, and potent second-generation type II FLT3 tyrosine kinase inhibitor. It inhibits wild-type FLT3 and FLT3-ITD autophosphorylation in MV4-11 cells and can induce apoptosis.
  • Orally active FLT3 tyrosine kinase inhibitor
  • Highly selective and potent
  • Inhibits FLT3 and FLT3-ITD autophosphorylation
  • Can be linked to VHL ligand for PROTAC FLT3 degrader formation
  • Induces apoptosis