Learn More
Medchemexpress LLC Quizartinib dihydrochloride | 1132827-21-4 | MFCD18633276 | 99.8% | C29H34Cl2N6O4S | 10MG
SDP
Catalog No. 5000259816
Encompass_Preferred
Quizartinib dihydrochloride is the dihydrochloride salt of quizartinib, a second-generation, type II FLT3 tyrosine kinase inhibitor used in research on FLT3-mutant acute myeloid leukemia. It is supplied as a white to off-white solid with high reported purity and is used in biochemical and cellular assays to study FLT3 signaling and inhibitor activity.
  • High purity suitable for research applications.
  • White to off-white solid form for straightforward handling.
  • Molecular formula C29H34Cl2N6O4S and molecular weight 633.59 g/mol for accurate calculations.
  • Stable when stored sealed and protected from moisture under recommended conditions.
  • Available in small pack sizes appropriate for preclinical studies.
Catalog No. 50-002-59816 Supplier Medchemexpress LLC Supplier No. HY1421710MG
Quizartinib dihydrochloride is the dihydrochloride salt of quizartinib, a second-generation, type II FLT3 tyrosine kinase inhibitor used in research on FLT3-mutant acute myeloid leukemia. It is supplied as a white to off-white solid with high reported purity and is used in biochemical and cellular assays to study FLT3 signaling and inhibitor activity.
  • High purity suitable for research applications.
  • White to off-white solid form for straightforward handling.
  • Molecular formula C29H34Cl2N6O4S and molecular weight 633.59 g/mol for accurate calculations.
  • Stable when stored sealed and protected from moisture under recommended conditions.
  • Available in small pack sizes appropriate for preclinical studies.