Learn More
Bioworld Quoirin & Lepoivre Medium, 50 L
Catalog No. 50254770
Encompass_Preferred
Quoirin & Lepoivre MediumContains Micronutrients and Macronutrients as described by Quoirin & Lepoivre (1977) and Quoirin et al. (1977)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 3.56 g/LpH: 3.5−4.5Soluble in water
Catalog No. 50-254-770 Supplier Bioworld Supplier No. 306301074
Add to Cart
Add to Cart
Quoirin & Lepoivre MediumContains Micronutrients and Macronutrients as described by Quoirin & Lepoivre (1977) and Quoirin et al. (1977)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 3.56 g/LpH: 3.5−4.5Soluble in water