Learn More
Bioworld Quoirin & Lepoivre Medium w/ Vitamins, 50 L
Catalog No. 50254773
Encompass_Preferred
Quoirin & Lepoivre MediumContains Macronutrients, Micronutrients, and Vitamins as described by Quoirin & Lepoivre (1977)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested
Catalog No. 50-254-773 Supplier Bioworld Supplier No. 306301084
Quoirin & Lepoivre MediumContains Macronutrients, Micronutrients, and Vitamins as described by Quoirin & Lepoivre (1977)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested