Learn More
Alkali Scientific Qwik-Snap™ 0.5mL Low Binding & Low Adhesion Microcentrifuge Tubes (0.65 capacity), Colorless, 500/Pack
SDP
Catalog No. 503026569
Encompass_Preferred
NoStick, Graduated Microcentrifuge Tubes. Boil-proof caps are ideal for rigorous high-temp applications and tight storage conditions. Crystal-clear polymer facilitates visualization of samples. Rated to 20,000 x G. Graduations at every 100 µL for easy volume estimation. Available with proprietary NoStickresin to ensure maximum sample recovery. Temperature range: -80˚C to +121˚C. Manufactured from high-quality, virgin polypropylene to ensure purity and provide excellent optical clarity. Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free. Cap Description:Boil-Proof Cap Color: -00 Clear Packaging: 500/unit, 10 units/case
Catalog No. 50-302-6569 Supplier Alkali Scientific Supplier No. C3023S
NoStick, Graduated Microcentrifuge Tubes. Boil-proof caps are ideal for rigorous high-temp applications and tight storage conditions. Crystal-clear polymer facilitates visualization of samples. Rated to 20,000 x G. Graduations at every 100 µL for easy volume estimation. Available with proprietary NoStickresin to ensure maximum sample recovery. Temperature range: -80˚C to +121˚C. Manufactured from high-quality, virgin polypropylene to ensure purity and provide excellent optical clarity. Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free. Cap Description:Boil-Proof Cap Color: -00 Clear Packaging: 500/unit, 10 units/case