Learn More
Alkali Scientific Qwik-Snap™ 0.6mL MicroTubes, Clear, Sterile (10 Pouches of 50 Tubes), 500/cs
SDP
Catalog No. 502335956
Encompass_Preferred
Qwik-Snap™ MicroTubes: Centrifugal Speed: 29,000 x g. Can withstand boiling up to 121℃ and freezing down to -80℃. Snaps into place for a secure seal. 100% polypropylene, RNase/DNase free. Clear, non-binding surface, graduated. Certified RNase, DNase and Pyrogen free. Pack of 500 clear, sterile 0.6mL centrifuge tubes.
Catalog No. 50-233-5956 Supplier Alkali Scientific Supplier No. C3008S
Qwik-Snap™ MicroTubes: Centrifugal Speed: 29,000 x g. Can withstand boiling up to 121℃ and freezing down to -80℃. Snaps into place for a secure seal. 100% polypropylene, RNase/DNase free. Clear, non-binding surface, graduated. Certified RNase, DNase and Pyrogen free. Pack of 500 clear, sterile 0.6mL centrifuge tubes.