Learn More
Alkali Scientific Qwik-Snap™ 1.7mL Low Binding & Low Adhesion Microcentrifuge Tubes, Colorless, 250/Pk
SDP
Catalog No. 503026471
Encompass_Preferred
Alkali Scientific Qwik-Snap MicroTubes, Low Binding & Low Adhesion are made of premium polypropylene plastic which allows for a crystal clear tube and high centrifugal strength. They feature a secure-snap cap that ensures a strong seal and eliminates leakage and evaporation. The 0.6mL tubes are packaged in bags of 1000 and the 1.7mL & 2mL tubes come 500 per pack. Available in 1.7mL (graduated to 1.5mL), 2.0mL, and 0.6mL sizes, the 1.7mL and 0.6mL tubes are also available in assorted colors. Capable of withstanding boiling temperatures up to 121ËC and freezing down to -80ËC, the Qwik-Snap micro centrifuge tubes are the perfect choice for your research. Centrifugal Speed: 29,000 x g Withstands boiling up to 121ËC and freezing down to -80ËC Snaps into place for a secure seal 100% polypropylene RNase/DNase free Clear, non-binding surface, graduated Certified RNase, DNase & Pyrogen free Sterile options (SAL 10-6) and assorted colors available
Catalog No. 50-302-6471 Supplier Alkali Scientific Supplier No. C3019S
Alkali Scientific Qwik-Snap MicroTubes, Low Binding & Low Adhesion are made of premium polypropylene plastic which allows for a crystal clear tube and high centrifugal strength. They feature a secure-snap cap that ensures a strong seal and eliminates leakage and evaporation. The 0.6mL tubes are packaged in bags of 1000 and the 1.7mL & 2mL tubes come 500 per pack. Available in 1.7mL (graduated to 1.5mL), 2.0mL, and 0.6mL sizes, the 1.7mL and 0.6mL tubes are also available in assorted colors. Capable of withstanding boiling temperatures up to 121ËC and freezing down to -80ËC, the Qwik-Snap micro centrifuge tubes are the perfect choice for your research. Centrifugal Speed: 29,000 x g Withstands boiling up to 121ËC and freezing down to -80ËC Snaps into place for a secure seal 100% polypropylene RNase/DNase free Clear, non-binding surface, graduated Certified RNase, DNase & Pyrogen free Sterile options (SAL 10-6) and assorted colors available