Learn More
Alkali Scientific Qwik-Snap™ 1.7mL Microcentrifuge Tubes, Opaque Black, Self Standing Bag, with 10 Cap Locks, 500/cs
SDP
Catalog No. 502336084
Encompass_Preferred
Qwik-Snap™ MicroTubes: Centrifugal Speed: 29,000 x g. Can withstand boiling up to 121℃ and freezing down to -80℃. Snaps into place for a secure seal. 100% polypropylene, RNase/DNase free. Clear, non-binding surface, graduated. Certified RNase, DNase and Pyrogen free. Pack of 500 opaque black 1.7mL centrifuge tubes.
Catalog No. 50-233-6084 Supplier Alkali Scientific Supplier No. C3017OB
Qwik-Snap™ MicroTubes: Centrifugal Speed: 29,000 x g. Can withstand boiling up to 121℃ and freezing down to -80℃. Snaps into place for a secure seal. 100% polypropylene, RNase/DNase free. Clear, non-binding surface, graduated. Certified RNase, DNase and Pyrogen free. Pack of 500 opaque black 1.7mL centrifuge tubes.