Learn More
Alkali Scientific Qwik-Snap™ 2.0mL MicroTubes, Low Binding and Low Adhesion, Natural, (200/pk) 2000/cs
SDP
Catalog No. 502336057
Encompass_Preferred
Qwik-Snap™ MicroTubes: Centrifugal Speed: 29,000 x g. Can withstand boiling up to 121℃ and freezing down to -80℃. Snaps into place for a secure seal. 100% polypropylene, RNase/DNase free. Clear, non-binding surface, graduated. Certified RNase, DNase and Pyrogen free. Pack of 1,000 clear 2.0mL centrifuge tubes.
Catalog No. 50-233-6057 Supplier Alkali Scientific Supplier No. C3021
Add to Cart
Add to Cart
Qwik-Snap™ MicroTubes: Centrifugal Speed: 29,000 x g. Can withstand boiling up to 121℃ and freezing down to -80℃. Snaps into place for a secure seal. 100% polypropylene, RNase/DNase free. Clear, non-binding surface, graduated. Certified RNase, DNase and Pyrogen free. Pack of 1,000 clear 2.0mL centrifuge tubes.