Learn More
Description
- Prepared by monoclonal immunochromatographic assay method
- Uses unpreserved stool specimen
- Easy-to-use lateral flow cassette
- Detects antibodies in10 min.,
- Built-in quality: Includes an internal control within the cassette and an external positive
- Used as a control reagent
- Operates without an equipment
- Room storage temperature at 15°C to 30°C
- The test contains
- 1 positive control reagent
- 20 sample preparation tubes with sampler attached to tube cap
- 20 test cassettes
Specifications
Specifications
| CE Marker | Yes |
| Clia Complexity | Moderate |
| Detectable Analytes | H. pylori Antigen |
| Type | Rapid Antigen Test |
| Disposable | Single-use |
| No. of Reactions | 20 Tests |
| Packaging Type | Cassette |
| Form | Ready-to-Use (RTU) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
