Learn More
MSC QwikProducts Coil Cleaner, pH Neutral, Microchannel Safe, Concentrated, 1 gal.
Catalog No. 5000505947
Encompass
QwikBright is a pH neutral cleaner/protectant specifically formulated for microchannel coils. It quickly penetrates and dissolves grease, dirt, grime, lint, dust, and bugs without removing existing protective film, while also adding its own protective film. It can be used indoors or outdoors on any type of heat exchanger coil.
  • Safe for microchannel heat exchangers
  • Neutral pH formulation
  • Tough on grease, dirt, and dust
  • Works on indoor and outdoor coils
  • Includes metal protectant to extend life
  • Heavy duty foaming action
  • Improves coil efficiency
  • Super concentrated (mix 1 part with 4 to 8 parts water)
Catalog No. 50-005-05947 Supplier MSC Supplier No. 26085837
QwikBright is a pH neutral cleaner/protectant specifically formulated for microchannel coils. It quickly penetrates and dissolves grease, dirt, grime, lint, dust, and bugs without removing existing protective film, while also adding its own protective film. It can be used indoors or outdoors on any type of heat exchanger coil.
  • Safe for microchannel heat exchangers
  • Neutral pH formulation
  • Tough on grease, dirt, and dust
  • Works on indoor and outdoor coils
  • Includes metal protectant to extend life
  • Heavy duty foaming action
  • Improves coil efficiency
  • Super concentrated (mix 1 part with 4 to 8 parts water)