Learn More
Abcam Q x -222, Na+ channel blocker, 10MG
Catalog No. 502707864
Encompass_Preferred
MW 256.77 Da, Purity >99%. Na+ channel blocker. Lidocaine derivative. Also available in simple stock solutions (ab146678) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7864 Supplier Abcam Supplier No. AB12012610MG
MW 256.77 Da, Purity >99%. Na+ channel blocker. Lidocaine derivative. Also available in simple stock solutions (ab146678) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement