Learn More
Electron Microscopy Sciences QuantomiX WetSEM QX -302 QX -302 Imaging Buffer
SDP
Catalog No. 5031356
Encompass_Preferred
QX-302 Imaging Buffer is a solution optimized for imaging samples in an SEM with QX-302 capsules and is formulated to minimize damage to the samples by the electron beam.
Catalog No. 50-313-56 Supplier Electron Microscopy Sciences Supplier No. IB74
QX-302 Imaging Buffer is a solution optimized for imaging samples in an SEM with QX-302 capsules and is formulated to minimize damage to the samples by the electron beam.