Learn More
Electron Microscopy Sciences QuantomiX WetSEM QX -302 Starter Kit
SDP
Catalog No. 5031340
Encompass_Preferred
The QX-302 Capsules are used for imaging various thick, non-adherent samples, such as tissue biopsies, plants and material specimens, in their natural wet state. The capsule is suitable for variable sample sizes, with a maximum diameter of 3 mm and thickness of up to 1 mm. No coating or embedding of the sample are required, enabling electron microscopy imaging with easy sample preparation comparable to light microscopy. Includes QX-302 Capsules and all the necessary accessories.
Catalog No. 50-313-40 Supplier Electron Microscopy Sciences Supplier No. SK30212
The QX-302 Capsules are used for imaging various thick, non-adherent samples, such as tissue biopsies, plants and material specimens, in their natural wet state. The capsule is suitable for variable sample sizes, with a maximum diameter of 3 mm and thickness of up to 1 mm. No coating or embedding of the sample are required, enabling electron microscopy imaging with easy sample preparation comparable to light microscopy. Includes QX-302 Capsules and all the necessary accessories.