Learn More
Medchemexpress LLC QX-314 (bromide) | 24003-58-5 | MFCD00083182 | 96.6% | 343.30 | C16H27BrN2O | 1 ML
SDP
Catalog No. 5000213181
Encompass_Preferred
QX-314 bromide is a membrane-impermeable, permanently charged sodium channel blocker provided as a ready-to-use 10 mM solution in DMSO or as a solid for reconstitution. It is commonly used in electrophysiology and neurophysiology experiments to selectively block intracellular voltage-gated sodium channels during recordings and manipulations.
  • Membrane-impermeable quaternary lidocaine derivative.
  • Available as 10 mM solution in DMSO or solid for reconstitution.
  • Used to block intracellular voltage-gated sodium channels in electrophysiology.
  • CAS number 24003-58-5 for unambiguous identification.
  • Supplied with reported purity suitable for research applications.
Catalog No. 50-002-13181 Supplier Medchemexpress LLC Supplier No. HY10135010MM 1ML
QX-314 bromide is a membrane-impermeable, permanently charged sodium channel blocker provided as a ready-to-use 10 mM solution in DMSO or as a solid for reconstitution. It is commonly used in electrophysiology and neurophysiology experiments to selectively block intracellular voltage-gated sodium channels during recordings and manipulations.
  • Membrane-impermeable quaternary lidocaine derivative.
  • Available as 10 mM solution in DMSO or solid for reconstitution.
  • Used to block intracellular voltage-gated sodium channels in electrophysiology.
  • CAS number 24003-58-5 for unambiguous identification.
  • Supplied with reported purity suitable for research applications.