Learn More
Selleck Chemical LLC QX-314 chloride
SDP
Catalog No. 5000003610
Encompass_Preferred
QX-314 chloride directly activates and permeates the human isoforms of TRPV1 and TRPA1 to induce inhibition of sodium channels
Catalog No. 50-000-03610 Supplier Selleck Chemical LLC Supplier No. S78485MG
QX-314 chloride directly activates and permeates the human isoforms of TRPV1 and TRPA1 to induce inhibition of sodium channels