Learn More
Medchemexpress LLC QX-314 chloride | 5369-03-9 | 100.0% | 298.85 | 100 MG
SDP
Catalog No. 5000346655
Encompass_Preferred
QX-314 chloride is a membrane-impermeable, permanently charged sodium channel blocker. It is intended for research use only.
  • Membrane-impermeable sodium channel blocker
  • Exerts biphasic effects on transient receptor potential vanilloid subtype 1 channels (TRPV1) in vitro
  • Activates TRPV1 in a concentration-dependent manner
  • Inhibits calcium currents in hippocampal CA1 pyramidal neurons intracellularly
Catalog No. 50-003-46655 Supplier Medchemexpress LLC Supplier No. HY10850510MM/1ML
QX-314 chloride is a membrane-impermeable, permanently charged sodium channel blocker. It is intended for research use only.
  • Membrane-impermeable sodium channel blocker
  • Exerts biphasic effects on transient receptor potential vanilloid subtype 1 channels (TRPV1) in vitro
  • Activates TRPV1 in a concentration-dependent manner
  • Inhibits calcium currents in hippocampal CA1 pyramidal neurons intracellularly