Learn More
Selleck Chemical LLC QX-314 chloride
SDP
Supplier:  Selleck Chemical LLC S784825MG
Encompass_Preferred
QX-314 chloride directly activates and permeates the human isoforms of TRPV1 and TRPA1 to induce inhibition of sodium channels
Catalog No. 50-000-03873