Learn More
Abcam Q x -314 chloride (N-Ethyllidocaine chloride) (mM/ml), Na+ channel blocker, 100MM_ML
Catalog No. 502705668
Encompass_Preferred
MW 298.8 Da, Purity >99%. A membrane impermeable quaternary lidocaine derivative. Blocks voltage-sensitive Na+ channels. Soluble in 1 ml water to give specified mM/ml concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5668 Supplier Abcam Supplier No. AB144493100MM_ML
MW 298.8 Da, Purity >99%. A membrane impermeable quaternary lidocaine derivative. Blocks voltage-sensitive Na+ channels. Soluble in 1 ml water to give specified mM/ml concentration.
The product is subject to the following: Abcam Restricted Use Statement