Learn More
Cayman Chemical QX-314 (chloride), Purity: ≥98%, CAS: 5369-03-9, 50 mg
SDP
Catalog No. 502967890
Encompass_Preferred
A membrane-impermeant lidocaine derivative that when combined with capsaicin selectively blocks sodium channels on nociceptive neurons via the TRPV1 channel; decreases the pain response without imparting numbness or paralysis associated with other local anesthetics
Catalog No. 50-296-7890 Supplier Cayman Chemical Supplier No. 1845450 MG
A membrane-impermeant lidocaine derivative that when combined with capsaicin selectively blocks sodium channels on nociceptive neurons via the TRPV1 channel; decreases the pain response without imparting numbness or paralysis associated with other local anesthetics