Learn More
Bio Rad Laboratories QX ONE DROPLET READR OIL 700ML
Catalog No. NC1879379
Encompass
  • Formulated for droplet analysis in digital PCR applications.
  • Supports seamless integration within the QX ONE ddPCR System.
  • Enables accurate reading and quantification of PCR-positive and PCR-negative droplets.
  • Critical for high precision and sensitivity in digital PCR workflows.
  • Volume provided is 700 ml.
  • Automated workflow assistance ensures reliable performance.
Catalog No. NC1879379 Supplier Bio Rad Laboratories Supplier No. 12006057
  • Formulated for droplet analysis in digital PCR applications.
  • Supports seamless integration within the QX ONE ddPCR System.
  • Enables accurate reading and quantification of PCR-positive and PCR-negative droplets.
  • Critical for high precision and sensitivity in digital PCR workflows.
  • Volume provided is 700 ml.
  • Automated workflow assistance ensures reliable performance.