Learn More
Cayman Chemical QX-314 (bromide), Purity: ≥98%, CAS: 24003-58-5, 50 mg
SDP
Catalog No. 502888512
Encompass_Preferred
A membrane-impermeant lidocaine derivative that when combined with capsaicin selectively blocks sodium channels on nociceptive neurons via the TRPV1 channel; decreases the pain response without imparting numbness or paralysis associated with other local anesthetics
Catalog No. 50-288-8512 Supplier Cayman Chemical Supplier No. 1001103250 MG
A membrane-impermeant lidocaine derivative that when combined with capsaicin selectively blocks sodium channels on nociceptive neurons via the TRPV1 channel; decreases the pain response without imparting numbness or paralysis associated with other local anesthetics