Learn More
ILEX LIFE SCIENCES LLC QuickZyme Human MMP-7 Activity Assay Kit, 96 Assays, 96-well plate format, immunoassay for active or total matrix metalloproteinase 7 (conditioned culture medium, serum, urine, tissue homogenates samples)
SDP
Catalog No. 502339459
Encompass_Preferred
The QuickZyme Human MMP-7 Activity Assay is an enzyme activity immunoassay that enables you to specifically measure both endogenous active MMP-7 and total active MMP-7 (following activation of pro-MMP-7 on the plate with APMA). It can be used for the measurement of MMP-7 activity in various biological samples, such as conditioned culture media, tissue homogenates, serum, plasma and urine. This 96-well plate format assay is based on the QuickZyme technology, using a modified pro-enzyme as a substrate, which upon activation is able to release color from a chromogenic peptide substrate. This multiplication step provides a unique assay sensitivity. ASSAY RANGE: 0.004 ¿ 16 ng/ml. SENSITIVITY: 30 pg/ml (3 h incubation); 4 pg/ml (25 h incubation). SYNONYMS: Matrix metalloproteinase-7, MMP7, Matrilysin, Matrilysin-1, Matrin, Pump-1 protease, Uterine metalloproteinase.
Catalog No. 50-233-9459 Supplier ILEX LIFE SCIENCES LLC Supplier No. QZBMMP7H
The QuickZyme Human MMP-7 Activity Assay is an enzyme activity immunoassay that enables you to specifically measure both endogenous active MMP-7 and total active MMP-7 (following activation of pro-MMP-7 on the plate with APMA). It can be used for the measurement of MMP-7 activity in various biological samples, such as conditioned culture media, tissue homogenates, serum, plasma and urine. This 96-well plate format assay is based on the QuickZyme technology, using a modified pro-enzyme as a substrate, which upon activation is able to release color from a chromogenic peptide substrate. This multiplication step provides a unique assay sensitivity. ASSAY RANGE: 0.004 ¿ 16 ng/ml. SENSITIVITY: 30 pg/ml (3 h incubation); 4 pg/ml (25 h incubation). SYNONYMS: Matrix metalloproteinase-7, MMP7, Matrilysin, Matrilysin-1, Matrin, Pump-1 protease, Uterine metalloproteinase.