Learn More
ILEX LIFE SCIENCES LLC QuickZyme Human MMP-8 Activity Assay Kit, 96 Assays, 96-well plate format, immunoassay for active or total matrix metalloproteinase 8 (cell culture conditioned medium, serum, plasma, urine, saliva, tissue homogenates samples)
SDP
Catalog No. 502339477
Encompass_Preferred
The QuickZyme Human MMP-8 Activity Assay is an enzyme activity immunoassay that enables you to specifically measure both endogenous active MMP-8 and total active MMP-8 (following activation of pro-MMP-8 on the plate with APMA). It can be used for the measurement of MMP-8 activity in various biological samples, such as cell culture conditioned medium, serum, plasma, urine, saliva and tissue homogenates. This 96-well plate format assay is based on the QuickZyme technology, using a modified pro-enzyme as a substrate, which upon activation is able to release color from a chromogenic peptide substrate. This multiplication step provides a unique assay sensitivity. ASSAY RANGE: 10 ¿ 24000 pg/ml. SENSITIVITY: 100 pg/ml (2 h incubation); 24 pg/ml (6 h incubation); 4 pg/ml (24 h incubation). SYNONYMS: Matrix metalloproteinase-8, MMP8, Neutrophil collagenase, PMNL collagenase, PMNL-CL.
Catalog No. 50-233-9477 Supplier ILEX LIFE SCIENCES LLC Supplier No. QZBMMP8H
The QuickZyme Human MMP-8 Activity Assay is an enzyme activity immunoassay that enables you to specifically measure both endogenous active MMP-8 and total active MMP-8 (following activation of pro-MMP-8 on the plate with APMA). It can be used for the measurement of MMP-8 activity in various biological samples, such as cell culture conditioned medium, serum, plasma, urine, saliva and tissue homogenates. This 96-well plate format assay is based on the QuickZyme technology, using a modified pro-enzyme as a substrate, which upon activation is able to release color from a chromogenic peptide substrate. This multiplication step provides a unique assay sensitivity. ASSAY RANGE: 10 ¿ 24000 pg/ml. SENSITIVITY: 100 pg/ml (2 h incubation); 24 pg/ml (6 h incubation); 4 pg/ml (24 h incubation). SYNONYMS: Matrix metalloproteinase-8, MMP8, Neutrophil collagenase, PMNL collagenase, PMNL-CL.