Learn More
ILEX LIFE SCIENCES LLC QuickZyme Hydroxyproline Assay Kit Colorimetric Method 1/EA
SDP
Catalog No. 502120032
Encompass_Preferred
QuickZyme Hydroxyproline Assay Kit 2 x 96 Assays (includes 2x 96-well plates) The QuickZyme Hydroxyproline Assay Kit enables quantitative collagen analysis in all types of tissues in a fast and simple 96-well plate format This colorimetric assay does not need the drying step following acid hydrolysis of collagen for which often special equipment is needed This assay is the ONLY hydroxyproline-based assay that does not need this laborious step resulting in a fast sensitive and easy assay NOTE This kit does not contain tubes or a protocol for collagen hydrolysis If you need these items please order the QuickZyme Total Collagen Assay Application conditioned media cell extracts tissues tissue extracts Sensitivity 2 4 M (0 3 g/ml) Assay-time 2 hours QuickZyme Hydroxyproline Assay can be paired with QuickZyme Total Protein Assay to allow the expression of data as hydroxyproline quantity per total amount of protein
Catalog No. 50-212-0032 Supplier ILEX LIFE SCIENCES LLC Supplier No. QZBHYPRO2
QuickZyme Hydroxyproline Assay Kit 2 x 96 Assays (includes 2x 96-well plates) The QuickZyme Hydroxyproline Assay Kit enables quantitative collagen analysis in all types of tissues in a fast and simple 96-well plate format This colorimetric assay does not need the drying step following acid hydrolysis of collagen for which often special equipment is needed This assay is the ONLY hydroxyproline-based assay that does not need this laborious step resulting in a fast sensitive and easy assay NOTE This kit does not contain tubes or a protocol for collagen hydrolysis If you need these items please order the QuickZyme Total Collagen Assay Application conditioned media cell extracts tissues tissue extracts Sensitivity 2 4 M (0 3 g/ml) Assay-time 2 hours QuickZyme Hydroxyproline Assay can be paired with QuickZyme Total Protein Assay to allow the expression of data as hydroxyproline quantity per total amount of protein