Learn More
ILEX LIFE SCIENCES LLC QuickZyme Hydroxyproline Assay Kit, 96 Assays, 96-well plate format, colorimetric assay for collagen analysis (conditioned media cells, cell extract, tissue, tissue extract samples)
SDP
Catalog No. 502120031
Encompass_Preferred
The QuickZyme Hydroxyproline Assay Kit enables quantitative collagen analysis in all types of tissues, in a fast and simple 96-well plate format. This colorimetric assay does not need the drying step following acid hydrolysis of collagen for which often special equipment is needed. This assay is the only hydroxyproline-based assay that does not need this laborious step, resulting in a fast, sensitive, and easy assay. NOTE: This kit does not contain tubes or a protocol for collagen hydrolysis. If you need these items please order the QuickZyme Total Collagen Assay. Application: conditioned media cells, cell extracts, tissues, tissue extracts. Sensitivity: 2.4 µM (0.3 µg/ml). Assay-time: < 2 hours. Collagen is one of the main components of extracellular matrix. Dysregulation in collagen production results in pathologies such as fibrosis (too much collagen), or osteoarthritis (too little collagen). Thus measurement of collagen production is important in many disease related studies.
Catalog No. 50-212-0031 Supplier ILEX LIFE SCIENCES LLC Supplier No. QZBHYPRO1
The QuickZyme Hydroxyproline Assay Kit enables quantitative collagen analysis in all types of tissues, in a fast and simple 96-well plate format. This colorimetric assay does not need the drying step following acid hydrolysis of collagen for which often special equipment is needed. This assay is the only hydroxyproline-based assay that does not need this laborious step, resulting in a fast, sensitive, and easy assay. NOTE: This kit does not contain tubes or a protocol for collagen hydrolysis. If you need these items please order the QuickZyme Total Collagen Assay. Application: conditioned media cells, cell extracts, tissues, tissue extracts. Sensitivity: 2.4 µM (0.3 µg/ml). Assay-time: < 2 hours. Collagen is one of the main components of extracellular matrix. Dysregulation in collagen production results in pathologies such as fibrosis (too much collagen), or osteoarthritis (too little collagen). Thus measurement of collagen production is important in many disease related studies.