Learn More
BioVendor, LLC QuickZyme Total Protein Assay Kit 2 X 96-Assays
SDP
Supplier:  BioVendor, LLC QZBTOTPROT2
Encompass
QuickZyme Total Protein Assay Kit, 2 X 96-Assays. Quantitative measurement of total protein in acid hydrolyzates of fresh, frozen, fixed and paraffin embedded tissues in a fast & simple 96-well plate format! The QuickZyme Total Collagen and Hydroxyproline assays are used to determine the amount of collagen in biological samples such as tissues, tissue extracts, cell extracts and culture media. For proper interpretation of the data obtained, these should be compared to some reference quantity. The QuickZyme Total Protein Assay offers a convenient solution in such cases, allowing easy expression of the collagen or hydroxyproline amounts per total amount of protein. The QuickZyme Total Protein assay is developed to measure protein in acid tissue and/or protein hydrolyzates in such a way that it can be performed directly on the hydrolyzate without a drying or neutralization step. Dynamic range: 0.04-3.00 mg/ml. Sensitivity: 0.007 mg/ml. Colorimetric read-out at 570 nm.
Catalog No. 50-206-9136