Learn More
Strem, An Ascensus Company CAS# 18531-94-7. 1g. R-(+)-1,1'-Bi-2-naphthol, 99% (R)-BINOL. MFCD00004068
SDP
Supplier:  Strem, An Ascensus Company 0810001G
Encompass
CAS# 18531-94-7. 1g. R-(+)-1,1'-Bi-2-naphthol, 99% (R)-BINOL. MFCD00004068. Molecular Weight: 286.33. Molecular Formula: HOC10H6-C10H6OH. Color/form: white to off-white pwdr. Strem# 08-1000. http://www.strem.com/catalog/v/08-1000/
Catalog No. 50-901-09206