Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-1,1'-Bi(2-naphthylamine), 97%

Catalog No. p-7028460
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH25785MD 250 mg
AAH2578503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH25785MD Supplier Thermo Scientific Chemicals Supplier No. H25785MD
Only null left
Add to Cart

CAS: 18741-85-0 | C20H16N2 | 284.36 g/mol

(R)-(+)-1,1'-Bi(2-naphthylamine) derivatives have been used for asymmetric hydrogenations, cyclopropanations and formation of chiral lactones. It can be used for the synthesis of sterically hindered binaphthalene-based monophosphane ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-1,1′-Bi(2-naphthylamine) derivatives have been used for asymmetric hydrogenations, cyclopropanations and formation of chiral lactones. It can be used for the synthesis of sterically hindered binaphthalene-based monophosphane ligands.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18741-85-0
Molecular Formula C20H16N2
Molecular Weight (g/mol) 284.36
MDL Number MFCD00145204
InChI Key DDAPSNKEOHDLKB-UHFFFAOYSA-N
Synonym 1,1'-binaphthalene-2,2'-diamine, r-1,1'-binaphthalene-2,2'-diamine, s---1,1'-binaphthyl-2,2'-diamine, 1,1'-binaphthyl-2,2'-diamine, s---2,2'-diamino-1,1'-binaphthalene, 2,2'-diamino-1,1'-dinaphthyl, 1,1'-bi 2-naphthylamine, 1,1'-bi 2-naphthalenylamine, binam, r-+-2,2'-diamino-1,1'-binaphthalene
PubChem CID 20571
IUPAC Name 1-(2-aminonaphthalen-1-yl)naphthalen-2-amine
SMILES NC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1N

Specifications

Melting Point 242°C to 243°C
Quantity 250 mg
Beilstein 5279621
Solubility Information Slightly soluble in water.
Optical Rotation +150° (c=1 in Pyridine)
Formula Weight 284.36
Percent Purity 97%
Chemical Name or Material (R)-(+)-1,1'-Bi(2-naphthylamine)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.