Learn More
Medchemexpress LLC (R)-1,1'-Binaphthyl-2,2'-diyl hydrogenphosphate | 39648-67-4 | 348.29 | 250 MG
SDP
Catalog No. 5000449965
Encompass_Preferred
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl Hydrogenphosphate is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also described as a chiral ligand used in hydrocarboxylation reactions, forming complexes with rhodium to mediate asymmetric dipolar cyclodition of diazo compounds.
  • Used as a biochemical reagent
  • Can be used as a biological material or organic compound for life science related research
  • Chiral ligand for hydrocarboxylation reactions
  • Forms complexes with rhodium to mediate asymmetric dipolar cyclodition of diazo compounds
  • Used to resolve racial amines
  • Palladium derivatives used in asymmetric hydrocarboxylations
  • Rhodium derivatives used in dipolar cycloditions
  • Used in chiral Brønsted acid catalyzed enantioselective Mannich reaction
Catalog No. 50-004-49965 Supplier Medchemexpress LLC Supplier No. 250MGHYW007978
(R)-(-)-1,1'-Binaphthyl-2,2'-diyl Hydrogenphosphate is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also described as a chiral ligand used in hydrocarboxylation reactions, forming complexes with rhodium to mediate asymmetric dipolar cyclodition of diazo compounds.
  • Used as a biochemical reagent
  • Can be used as a biological material or organic compound for life science related research
  • Chiral ligand for hydrocarboxylation reactions
  • Forms complexes with rhodium to mediate asymmetric dipolar cyclodition of diazo compounds
  • Used to resolve racial amines
  • Palladium derivatives used in asymmetric hydrocarboxylations
  • Rhodium derivatives used in dipolar cycloditions
  • Used in chiral Brønsted acid catalyzed enantioselective Mannich reaction