Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-1-(1-Naphthyl)ethyl Isocyanate 90.0+%, TCI America™
SDP

Catalog No. I03365G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03365G 5 g
I03361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03365G Supplier TCI America Supplier No. I03365G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42340-98-7
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.24
MDL Number MFCD00004015
InChI Key GONOHGQPZFXJOJ-SNVBAGLBSA-N
Synonym r---1-1-naphthyl ethyl isocyanate, isocyanic acid r---1-1-naphthyl ethyl ester, 1-1r-1-isocyanatoethyl naphthalene, naphthalene,1-1r-1-isocyanatoethyl, 1r-1-naphthylethanisocyanate, 1-1-isocyanatoethyl naphthalene #, pubchem8100, r-nei, r-1-1-naphthyl ethyl isocyanate
PubChem CID 170633
IUPAC Name 1-[(1R)-1-isocyanatoethyl]naphthalene
SMILES C[C@@H](N=C=O)C1=C2C=CC=CC2=CC=C1

Specifications

Color Yellow
Quantity 5 g
UN Number 2206
Formula Weight 197.24
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material (R)-(-)-1-(1-Naphthyl)ethyl Isocyanate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.