Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-1,2-Diaminopropane Dihydrochloride 97.0+%, TCI America™
SDP

Catalog No. D28271G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D28271G Supplier TCI America Supplier No. D28271G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19777-67-4
Molecular Formula C3H12Cl2N2
Molecular Weight (g/mol) 147.04
MDL Number MFCD00239481
InChI Key AEIAMRMQKCPGJR-UHFFFAOYNA-N
Synonym (R)-1,2-Propanediamine Dihydrochloride, (R)-Propylenediamine Dihydrochloride
PubChem CID 12217444
IUPAC Name propane-1,2-diamine dihydrochloride
SMILES Cl.Cl.CC(N)CN

Specifications

Melting Point 242°C
Color White
Quantity 1 g
Formula Weight 147.04
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material (R)-1,2-Diaminopropane Dihydrochloride
EINECSNumber (2)-0149
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.