Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-1-(2-Naphthyl)ethanol 98.0+%, TCI America™
SDP

Catalog No. N07841G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N07841G Supplier TCI America Supplier No. N07841G

Chemical Identifiers

CAS 52193-85-8
Molecular Formula C12H12O
Molecular Weight (g/mol) 172.227
MDL Number MFCD00075344
InChI Key AXRKCRWZRKETCK-SECBINFHSA-N
Synonym (R)-(+)-alpha-Methyl-2-naphthalenemethanol, (R)-(+)-2-(1-Hydroxyethyl)naphthalene
PubChem CID 6976569
IUPAC Name (1R)-1-naphthalen-2-ylethanol
SMILES CC(C1=CC2=CC=CC=C2C=C1)O

Specifications

Melting Point 72°C
Color White
Quantity 1 g
Formula Weight 172.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-1-(2-Naphthyl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.