Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-1-(2-Pyrrolidinylmethyl)pyrrolidine 98.0+%, TCI America™
SDP

Catalog No. P19251G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P19251G Supplier TCI America Supplier No. P19251G

Chemical Identifiers

CAS 60419-23-0
Molecular Formula C9H20N2
Molecular Weight (g/mol) 156.27
MDL Number MFCD04117879
InChI Key YLBWRMSQRFEIEB-SECBINFHSA-P
PubChem CID 853151
IUPAC Name 1-{[(2R)-pyrrolidin-1-ium-2-yl]methyl}pyrrolidin-1-ium
SMILES C([C@H]1CCC[NH2+]1)[NH+]1CCCC1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 154.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (R)-(-)-1-(2-Pyrrolidinylmethyl)pyrrolidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.