Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-1-(3-(4-amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl)piperidin-1-yl)-2-(dimethylam | 1839150-56-9 | 98.5% | 471.55 | C26H29N7O2 | 5 MG
SDP

Catalog No. 5000264706
Encompass_Preferred

Selective FLT3 kinase inhibitor for laboratory research. Potent inhibitor (IC50 = 40 nM) used in preclinical studies to probe FLT3-driven signaling and apoptosis. Supplied as a high-purity small molecule with supporting datasheet, COA, and SDS; not for human use.

  • Potent FLT3 inhibition (IC50 40 nM).
  • High purity suitable for research (≈98.5%).
  • Molecular formula C26H29N7O2; molecular weight 471.55.
  • Soluble in DMSO for biological assays.
  • Available in multiple small-scale pack sizes for preclinical work.
  • Provided with datasheet, certificate of analysis, and safety data sheet.

Catalog No. 50-002-64706 Supplier Medchemexpress LLC Supplier No. HY1102935MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selective FLT3 kinase inhibitor for laboratory research. Potent inhibitor (IC50 = 40 nM) used in preclinical studies to probe FLT3-driven signaling and apoptosis. Supplied as a high-purity small molecule with supporting datasheet, COA, and SDS; not for human use.

  • Potent FLT3 inhibition (IC50 40 nM).
  • High purity suitable for research (≈98.5%).
  • Molecular formula C26H29N7O2; molecular weight 471.55.
  • Soluble in DMSO for biological assays.
  • Available in multiple small-scale pack sizes for preclinical work.
  • Provided with datasheet, certificate of analysis, and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.