Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol, 98%

Catalog No. AAH6089306
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6089306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6089306 Supplier Thermo Scientific Chemicals Supplier No. H6089306

missing translation for 'validMainframeMsg'

CAS: 127852-28-2 | C10H8F6O | 258.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127852-28-2
Molecular Formula C10H8F6O
Molecular Weight (g/mol) 258.16
MDL Number MFCD03093010
InChI Key MMSCIQKQJVBPIR-UHFFFAOYNA-N
Synonym r-1-3,5-bis trifluoromethyl phenyl ethanol, 1r-1-3,5-bis trifluoromethyl phenyl ethanol, r-1-3,5-bis-trifluoromethyl-phenyl-ethanol, r-1-3,5-bis-trifluoromethylphenyl ethanol, r-1-3,5-bis trifluoromethyl phenyl ethan-1-ol, benzenemethanol, a-methyl-3,5-bis trifluoromethyl-, ar, zlchem 1281
PubChem CID 2779031
IUPAC Name (1R)-1-[3,5-bis(trifluoromethyl)phenyl]ethanol
SMILES CC(O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 53°C to 58°C
Color White
Quantity 5 g
Formula Weight 258.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.