Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(-)-1,3-Butanediol 98%, Thermo Scientific™

Catalog No. AC217960010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC217960010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC217960010 Supplier Thermo Scientific Chemicals Supplier No. 217960010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6290-03-5
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
InChI Key PUPZLCDOIYMWBV-UHFFFAOYNA-N
Synonym r---1,3-butanediol, r-butane-1,3-diol, 3r-butane-1,3-diol, r-1,3-butanediol, 1,3-butanediol, 3r, r---1,3-butylene glycol, r---butane-1,3-diol, 1,3-butanediol, r
PubChem CID 637497
ChEBI CHEBI:52687
IUPAC Name (3R)-butane-1,3-diol
SMILES CC(CCO)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Refractive Index 1.4385 to 1.4405 (20°C, 589nm)
Linear Formula CH3CH(OH)CH2CH2OH
Quantity 1 g
Fieser 15,51
Specific Rotation -30° to -32° (20°C, 589nm) (c=1, C2H5OH)
Formula Weight 90.12
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material (R)-(-)-1, 3-Butanediol, 98%
Show More Show Less
Hazard Category HYGROSCOPIC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.