Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-1-(4-Bromophenyl)ethylamine, ChiPros, 99%, ee 98%

Catalog No. AAL2007914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2007914 25 g
AAL2007903 1 g
AAL2007906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL2007914 Supplier Thermo Scientific Chemicals Supplier No. L2007914
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 45791-36-4 | C8H10BrN | 200.079 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 45791-36-4
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.079
MDL Number MFCD00066025
InChI Key SOZMSEPDYJGBEK-ZCFIWIBFSA-N
Synonym r-1-4-bromophenyl ethylamine, r-+-1-4-bromophenyl ethylamine, r-1-4-bromophenyl ethanamine, 1r-1-4-bromophenyl ethanamine, 1r-1-4-bromophenyl ethan-1-amine, r-1-4-bromophenyl-ethylamine, r-4-bromo-alpha-methylbenzylamine, r-+-4-bromo a-methylbenzylamine, p-bromo-a-methylbenzylamine, pubchem14234
PubChem CID 853000
IUPAC Name (1R)-1-(4-bromophenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)Br)N

Specifications

Melting Point -25°C (neat)
Density 1.371
Boiling Point 140°C to 145°C (30 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.567
Quantity 25 g
UN Number UN2735
Beilstein 2412319
Sensitivity Air Sensitive
Formula Weight 200.08
Percent Purity 99%
Assay ee 98%
Chemical Name or Material (R)-(+)-1-(4-Bromophenyl)ethylamine
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 256-245-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.