Learn More
Medchemexpress LLC (R)-1-(4-chloro-2-oxo-1,2-dihydropyridin-3-yl)-N-(4-methyl-3-(N-(3-pyridinylmethyl)sulfamoyl)phenyl)-2-pyrrolidinecarboxamide | 2760881-74-9 | 98.0% | C21H22ClN5O4S | 10MG
SDP
Catalog No. 5000260209
Encompass_Preferred
BRD0639 is a first-in-class inhibitor of the PRMT5-substrate adaptor interaction that acts as a PRMT5 binding motif (PBM)-competitive agent for studying PBM-dependent PRMT5 activities. Supplied as a solid with analytical characterization, it is intended for research use in biochemical and cellular assays.
  • PBM-competitive mechanism enables selective modulation of PRMT5 interactions.
  • High reported purity appropriate for biochemical and cellular studies.
  • Solid form with recommended cold storage for extended stability.
  • Available in small research-scale packaging to support exploratory work.
  • Documented chemical identifiers and characterization for traceability.
Catalog No. 50-002-60209 Supplier Medchemexpress LLC Supplier No. HY13230910MG
BRD0639 is a first-in-class inhibitor of the PRMT5-substrate adaptor interaction that acts as a PRMT5 binding motif (PBM)-competitive agent for studying PBM-dependent PRMT5 activities. Supplied as a solid with analytical characterization, it is intended for research use in biochemical and cellular assays.
  • PBM-competitive mechanism enables selective modulation of PRMT5 interactions.
  • High reported purity appropriate for biochemical and cellular studies.
  • Solid form with recommended cold storage for extended stability.
  • Available in small research-scale packaging to support exploratory work.
  • Documented chemical identifiers and characterization for traceability.