Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-(+)-1-(4-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 99+%, Thermo Scientific™

Catalog No. AAL1632114
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1632114 25 g
AAL1632103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1632114 Supplier Thermo Scientific Chemicals Supplier No. L1632114

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(R)-(+)-1-(4-Methoxyphenyl)ethylamine use in diastereo-and enantioselective Michael addition reactions. Thiazoles has been prepared from (R)-(+)-1-(4-Methoxyphenyl)ethylamine.

Solubility
Soluble in water (10g/L).

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22038-86-4
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.209
MDL Number MFCD00671659
InChI Key JTDGKQNNPKXKII-SSDOTTSWSA-N
Synonym r-+-1-4-methoxyphenyl ethylamine, r-1-4-methoxyphenyl ethylamine, r-1-4-methoxyphenyl ethanamine, 1r-1-4-methoxyphenyl ethanamine, r-+-4-methoxy-alpha-methylbenzylamine, s-4-methoxy-alpha-methylbenzylamine, r +-1-4-methylphenyl ethylamine, pubchem9920, ksc496q8b, rm+-1-4-methoxyphenyl ethylamine
PubChem CID 641449
IUPAC Name (1R)-1-(4-methoxyphenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)OC)N

Specifications

Melting Point <-20°C
Density 1.02
pH 11
Boiling Point 65°C (0.38 mmHg)
Odor Amine-like
Refractive Index 1.533
Quantity 25 g
UN Number UN2735
Beilstein 2413029
Sensitivity Air Sensitive
Solubility Information Soluble in water (10g/L).
Optical Rotation +34° (Neat)
Formula Weight 151.21
Percent Purity ≥99%
Assay ee ≥99%
Chemical Name or Material (R)-(+)-1-(4-Methoxyphenyl)ethylamine
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 606-907-3
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.