Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (R)-1-Amino-2-propanol | 2799-16-8 | 75.11 | 1 ML
SDP

Catalog No. 5000447662
Encompass_Preferred

(R)-1-Amino-2-propanol, also known as (R)-1-aminopropan-2-ol, is a biochemical reagent that serves as a biological material or organic compound for life science-related research. It is specifically noted as a useful research intermediate for the synthesis of other bioactive small molecules with pharmaceutical applications.

  • Biochemical reagent: can be utilized as a biological material or organic compound.
  • Research intermediate: useful in the synthesis of other bioactive small molecules with pharmaceutical applications.
  • Available in various quantities: offered in solid form (10g, 25g, 50g) and as a 10 mM solution in DMSO.
  • High purity: available with a purity of 99.95%.

Catalog No. 50-004-47662 Supplier Medchemexpress LLC Supplier No. HYW00229510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(R)-1-Amino-2-propanol, also known as (R)-1-aminopropan-2-ol, is a biochemical reagent that serves as a biological material or organic compound for life science-related research. It is specifically noted as a useful research intermediate for the synthesis of other bioactive small molecules with pharmaceutical applications.

  • Biochemical reagent: can be utilized as a biological material or organic compound.
  • Research intermediate: useful in the synthesis of other bioactive small molecules with pharmaceutical applications.
  • Available in various quantities: offered in solid form (10g, 25g, 50g) and as a 10 mM solution in DMSO.
  • High purity: available with a purity of 99.95%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.