Learn More
Medchemexpress LLC R -1-Amino-3-chloro 10g
SDP
Catalog No. 5000206709
Encompass_Preferred
(R)-1-Amino-3-chloropropan-2-ol hydrochloride is a derivative of 3-chloro-l 2 propanediol ( -chlorohydrin) 3-chloro-l 2 propanediol (a-chlorohydrin) is a male chemosterilant[1]
Catalog No. 50-002-06709 Supplier Medchemexpress LLC Supplier No. HYW10864910G
(R)-1-Amino-3-chloropropan-2-ol hydrochloride is a derivative of 3-chloro-l 2 propanediol ( -chlorohydrin) 3-chloro-l 2 propanediol (a-chlorohydrin) is a male chemosterilant[1]